Learn More
Equilin 3-O-beta-D-Glucuronide Sodium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Equilin 3-O-beta-D-Glucuronide Sodium Salt |
| CAS | 27610-12-4 |
| Molecular Formula | C24H27NaO8 |
| Quantity | 1 mg |
| Synonym | 17-Oxoestra-1,3,5(10),7-tetraen-3-yl β-D-Glucopyranosiduronic Acid Monosodium Salt |
| InChI Formula | InChI=1S/C24H28O8.Na/c1-24-9-8-14-13-5-3-12(10-11(13)2-4-15(14)16(24)6-7-17(24)25)31-23-20(28)18(26)19(27)21(32-23)22(29)30;/h3-5,10,14,16,18-21,23,26-28H,2,6-9H2,1H3,(H,29,30);/q;+1/p-1/t14-,16+,18+,19+,20-,21+,23-,24+;/m1./s1 |
| SMILES | [Na+].C[C@]12CC[C@H]3C(=CCc4cc(O[C@@H]5O[C@@H]([C@@H](O)[C@H](O)[C@H]5O)C(=O)[O-])ccc34)[C@@H]1CCC2=O |
| Molecular Weight (g/mol) | 466.46 |
| Formula Weight | 466.1604 |