missing translation for 'onlineSavingsMsg'
Learn More

EPZ020411 hydrochloride, MedChemExpress

Product Code. 16498167
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16498167 5 mg 5mg
16486046 100 mg 100mg
16455458 10 mg 10mg
16421637 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16498167 Supplier Medchem Express Supplier No. HY12970A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress EPZ020411 hydrochloride is a selective inhibitor of PRMT6 with an IC50 of 10 nM, it has >10 folds selectivity for PRMT6 over PRMT1 and PRMT8. EPZ020411 hydrochloride can be used for the research of cancer.

EPZ020411 hydrochloride is a selective inhibitor of PRMT6 with an IC50 of 10 nM, it has >10 folds selectivity for PRMT6 over PRMT1 and PRMT8. EPZ020411 hydrochloride can be used for the research of cancer.

Specifications

Chemical Name or Material EPZ020411 hydrochloride
CAS 2070015-25-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C25H39ClN4O3
Quantity 5 mg
Solubility Information DMSO : 50 mg/mL (104.37 mM; Need ultrasonic) ∣H2O : 20 mg/mL (41.75 mM; ultrasonic and warming and heat to 60°C)
SMILES CNCCN(C)CC1=CNN=C1C2=CC=C(O[C@H]3C[C@H](OCCC4CCOCC4)C3)C=C2.Cl[H]
Molecular Weight (g/mol) 479.06
Formula Weight 479.06
Percent Purity 97.21%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.