missing translation for 'onlineSavingsMsg'
Learn More

EPZ005687, MedChemExpress

Product Code. 16496618
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16496618

Brand: Medchem Express HY15555

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress EPZ005687 is a potent and selective inhibitor of EZH2 with Ki of 24 nM, and has 50-fold selectivity against EZH1 and 500-fold selectivity against 15 other protein methyltransferases.

EPZ005687 is a potent and selective inhibitor of EZH2 with Ki of 24 nM, and has 50-fold selectivity against EZH1 and 500-fold selectivity against 15 other protein methyltransferases.

Specifications

Chemical Name or Material EPZ005687
Physical Form Solid
CAS 1396772-26-1
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C32H37N5O3
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 9.4 mg/mL (17.42 mM; Need ultrasonic and warming)
SMILES O=C(NCC1=C(C)C=C(C)NC1=O)C2=CC(C3=CC=C(CN4CCOCC4)C=C3)=CC5=C2C=NN5C6CCCC6
Molecular Weight (g/mol) 539.67
Formula Weight 539.67
Percent Purity 95.11%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.