missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Epsin 1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599300
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total EPN1.
This gene encodes a member of the epsin protein family. The encoded protein binds clathrin and is involved in the endocytosis of clathrin-coated vesicles. Loss of function of this gene is associated with reduced tumor growth and progression in certain cancer types.
Specifications
| Epsin 1 | |
| Polyclonal | |
| Unconjugated | |
| EPN1 | |
| AA269831; EH domain-binding mitotic phosphoprotein; Epn1; EPS-15-interacting protein 1; Epsin 1; epsin-1; hypothetical protein LOC508199; Ibp1; Intersectin-EH-binding protein 1; LOW QUALITY PROTEIN: epsin-1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 117277, 13854, 29924 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O88339, Q80VP1, Q9Y6I3 | |
| EPN1 | |
| A synthesized peptide derived from human EPN1(Accession Q9Y6I3), corresponding to amino acid residues L140-L190. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction