Learn More
epsilon-Caprolactam-d10, CDN
Product Code. 30319468
Change view
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30319468 0.1 g 0.1g
30313388 0.05 g 0.05g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30319468 Supplier CDN Supplier No. CDND52530.1G
Please to purchase this item. Need a web account? Register with us today!
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material eta-Caprolactam-d10
CAS 169297-53-4
Molecular Formula C6HD10NO
Quantity 0.1 g
Synonym Hexahydro-3,4,5,6,7-d5-2 H-azepin-2-one-3,4,5,6,7-d5
InChI Formula InChI=1 S/C6H11NO/c8-6-4-2-1-3-5-7-6/h1-5H2,(H,7,8)/i1D2,2D2,3D2,4D2,5D2
SMILES [2 H]C1([2 H])NC(=O)C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C1([2 H])[2 H]
IUPAC Name 3,3,4,4,5,5,6,6,7,7-decadeuterioazepan-2-one
Molecular Weight (g/mol) 123.22
Formula Weight 123.1468 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity