Learn More
Eprobemide, MedChemExpress
Product Code. 16498698
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
Product Code. 16498698
Brand: Medchem Express HYB1413
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Eprobemide is a non-competitive reversible inhibitor of monoamine oxidase A.
Eprobemide is a non-competitive reversible inhibitor of monoamine oxidase A.
Specifications
Chemical Name or Material Eprobemide
CAS 87940-60-1
Color White
Purity Grade Notes Research
Molecular Formula C14H19ClN2O2
Quantity 10 mg
Synonym LIS 630
Solubility Information DMSO : ≥ 125 mg/mL (442.06 mM)
SMILES O=C(NCCCN1CCOCC1)C2=CC=C(Cl)C=C2
Molecular Weight (g/mol) 282.77
Formula Weight 282.77
Percent Purity 99.63%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation