missing translation for 'onlineSavingsMsg'
Learn More

National Lab Eppi™45 Kryo Boxes

Product Code. 15108588
Change view
Click to view available options
Color:
Neon Green
Neon Orange
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
15108588 Neon Green Each
15168588 Neon Orange -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15108588 Supplier National Lab Supplier No. KEA45V81NG
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Color Neon Green
Shape Rectangular
Holds 81 Vials up to 12.5 mm Diameter
Product Type Kryo Box
Dimensions (L x W x H) 130 x 130 mm
Length (Metric) 130 mm
Width (Metric) 130 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.