missing translation for 'onlineSavingsMsg'
Learn More

National Lab Eppi™45 Kryo Boxes

Product Code. 15168588
Click to view available options
Color:
Neon Green
Neon Orange
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15168588

Brand: National Lab KEA45V81NO

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Color Neon Orange
Shape Rectangular
Holds 81 Vials up to 12.5 mm Diameter
Product Type Kryo Box
Dimensions (L x W x H) 130 x 130 mm
Length (Metric) 130 mm
Width (Metric) 130 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.