Learn More
Eppendorf Eppendorf twin.tec™ PCR plate LoBind 384, skirted
Made from polypropylene, Eppendorf twin.tec PCR Plates LoBind enable you to recover nearly 100 % of your DNA/RNA molecules, and as there’s no surface coating, there’s no risk of sample contamination. Batch-tested and certified free from DNA, DNase, RNase and PCR inhibitors, they are perfect for PCR.
Brand:  Eppendorf 0030129687
Additional Details : Weight : 0.85000kg
Product Code. 16060582
  • 310.00 € / Pack of 300
Estimated Shipment: 24-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description
Description
Eppendorf twin.tec™ PCR plate LoBind 384, skirted
Specifications
Specifications
25