missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies EpiCult™ Plus Medium

Product Code. 17149231
Encompass
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17149231 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17149231 Supplier STEMCELL Technologies Supplier No. 06070

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For culture of human, mouse, and rat epithelial stem cells

  • EpiCult™ Plus Medium is a serum- and BPE-free culture medium for the expansion of epithelial stem cells from mammary, prostate, and other stratified and pseudostratified epithelial tissues from human, mouse, and rat
  • Epithelial cells can be expanded in EpiCult™ Plus Medium for multiple passages and for > 30 population doublings; for some human mammary cells, addition of fetal bovine serum may be required
  • EpiCult™ Plus Medium is not recommended for human keratinocytes or primary human airway cells; for culturing primary human airway cells, we recommend PneumaCult™-Ex Plus Medium (Catalog #05040)
  • Note: Hydrocortisone Stock Solution (Catalog #07925) is required for preparation of complete EpiCult™ Plus Medium

Specifications

Cell Type Epithelial Cells
For Use With (Application) Cell Culture
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.