missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EphA7 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98316H
This item is not returnable.
View return policy
Description
EphA7 Polyclonal specifically detects EphA7 in Mouse samples. It is validated for ELISA.
Specifications
| EphA7 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| ELISA | |
| HRP | |
| EC 2.7.10, EC 2.7.10.1, EHK3, EHK-3, EK11, EPH homology kinase 3, Eph homology kinase-3, EPH receptor A7, EphA7, EPH-like kinase 11, ephrin type-A receptor 7, HEK11, receptor protein-tyrosine kinase HEK11, tyrosine-protein kinase receptor EHK-3 | |
| Produced in rabbits immunized with purified, recombinant EphA7 (Q61772-1; Met 1-Ile 556). | |
| 0.1 mL | |
| Neuronal Cell Markers, Neuroscience, Neurotransmission, Protein Kinase | |
| 2045 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction