missing translation for 'onlineSavingsMsg'
Learn More

Epacadostat, MedChemExpress

Product Code. p-200100216
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
1 g
2 g
Unit Size:
100mg
10mg
1g
200mg
2g
500mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16491539

Brand: Medchem Express HY15689

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Epacadostat (INCB 024360) is a potent and selective indoleamine 2,3-dioxigenase 1 (IDO1) inhibitor with an IC50 of 71.8 nM.

Epacadostat (INCB 024360) is a potent and selective indoleamine 2,3-dioxigenase 1 (IDO1) inhibitor with an IC50 of 71.8 nM.

Specifications

Chemical Name or Material Epacadostat
CAS 1204669-58-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C11H13BrFN7O4S
Quantity 1 g
Synonym INCB 024360
Solubility Information DMSO : 100 mg/mL (228.19 mM; Need ultrasonic)
SMILES BrC1=C(F)C=CC(N/C(C2=NON=C2NCCNS(N)(=O)=O)=N\O)=C1
Molecular Weight (g/mol) 438.23
Formula Weight 438.23
Percent Purity 98.01%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.