missing translation for 'onlineSavingsMsg'
Learn More

Enzalutamide, MedChemExpress

Product Code. 16494067
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
1 g
2 g
Unit Size:
100mg
10mg
1g
200mg
2g
500mg
50mg
5mg
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16494067 50 mg 50mg
16480356 2 g 2g
16461946 500 mg 500mg
16451336 200 mg 200mg
16450618 5 mg 5mg
16428168 10 mg 10mg
16451959 1 g 1g
16407926 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 16494067 Supplier Medchem Express Supplier No. HY70002

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Enzalutamide (MDV3100) is an androgen receptor (AR) antagonist with an IC50 of 36 nM in LNCaP prostate cells. Enzalutamide is an autophagy activator.

Enzalutamide (MDV3100) is an androgen receptor (AR) antagonist with an IC50 of 36 nM in LNCaP prostate cells. Enzalutamide is an autophagy activator.

Specifications

Chemical Name or Material Enzalutamide
CAS 915087-33-1
Color White
Purity Grade Notes Research
Molecular Formula C21H16F4N4O2S
Quantity 50 mg
Synonym MDV3100
Solubility Information DMSO : ≥ 50 mg/mL (107.66 mM) ∣H2O : ≥ 0.1 mg/mL (0.22 mM)
SMILES S=C(N(C(C1(C)C)=O)C2=CC=C(C#N)C(C(F)(F)F)=C2)N1C3=CC(F)=C(C(NC)=O)C=C3
Molecular Weight (g/mol) 464.44
Formula Weight 464.44
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.