missing translation for 'onlineSavingsMsg'
Learn More

Memmert™ Entry Port 23 mm, Flap Closure, Custom Tbc Position In Right Side For U1060

Use these entry ports with convection ovens. Memmert™ Convection and Universal Oven Entry Points can be closed by flap in special positions at the back.

Brand:  Memmert™ U1060F5

Additional Details : Weight : 0.01100kg

Product Code. 16386067

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • Entry port with clear diameter
  • For Memmert Ovens
Specifications

Specifications

Models 30, 55, 75, 110, 160, 260, 450, 750, 1060
Stainless Steel
23 mm
Entry Port
0.9 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions