Learn More
Entecavir Monohydrate, TRC
Specifications
Specifications
| Chemical Name or Material | Entecavir Monohydrate |
| CAS | 209216-23-9 |
| Molecular Formula | C12 H15 N5 O3 . H2 O |
| Quantity | 1 mg |
| Synonym | 6H-Purin-6-one, 2-amino-1,9-dihydro-9-[(1S,3R,4S)-4-hydroxy-3-(hydroxymethyl)-2-methylenecyclopentyl]-, hydrate (1:1), 6H-Purin-6-one, 2-amino-1,9-dihydro-9-[(1S,3R,4S)-4-hydroxy-3-(hydroxymethyl)-2-methylenecyclopentyl]-, monohydrate (9CI), Entecavir monohydrate |
| InChI Formula | InChI=1S/C12H15N5O3.H2O/c1-5-6(3-18)8(19)2-7(5)17-4-14-9-10(17)15-12(13)16-11(9)20;/h4,6-8,18-19H,1-3H2,(H3,13,15,16,20);1H2/t6-,7-,8-;/m0./s1 |
| SMILES | O.NC1=Nc2c(ncn2[C@H]3C[C@H](O)[C@@H](CO)C3=C)C(=O)N1 |
| IUPAC Name | 2-amino-9-[(1S,3R,4S)-4-hydroxy-3-(hydroxymethyl)-2-methylidenecyclopentyl]-1H-purin-6-one;hydrate |
| Molecular Weight (g/mol) | 295.29 |
| Formula Weight | 295.1281 |