missing translation for 'onlineSavingsMsg'
Learn More

Entacapone, MedChemExpress

Product Code. p-200099396
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16486018

Brand: Medchem Express HY14280

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Entacapone is a potent, reversible, peripherally acting and orally active catechol-O-methyltransferase (COMT) inhibitor.

Entacapone inhibits COMT from rat brain, erythrocytes and liver with IC50 values of 10 nM, 20 nM, and 160 nM, respectively. Entacapone is selective for COMT over other catecholamine metabolizing enzymes, including MAO-A, MAO-B, phenolsulphotransferase M (PST-M) and PST-P (IC50s>50 μM). Entacapone can be used for the research of Parkinson's disease. Entacapone serves as a inhibitor of FTO demethylation with an IC50 of 3.5 μM, can be used for the research of metabolic disorders.

Specifications

Chemical Name or Material Entacapone
CAS 130929-57-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C14H15N3O5
Quantity 10 mg
Solubility Information DMSO : 33.33 mg/mL (109.17 mM; Need ultrasonic) ∣H2O : 2 mg/mL (6.55 mM; ultrasonic and adjust pH to 10 with NaOH)
SMILES O=C(N(CC)CC)/C(C#N)=C/C1=CC([N+]([O-])=O)=C(O)C(O)=C1
Molecular Weight (g/mol) 305.29
Formula Weight 305.29
Percent Purity 98.79%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.