missing translation for 'onlineSavingsMsg'
Learn More

Enoxacin hydrate, MedChemExpress

Product Code. 16402086
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16402086 500 mg 500mg
16428206 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16402086 Supplier Medchem Express Supplier No. HYB0268A
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Enoxacin hydrate (Enoxacin sesquihydrate), a fluoroquinolone, interferes with DNA replication and inhibits bacterial DNA gyrase (IC50=126 μg/ml) and topoisomerase IV (IC50=26.5 μg/ml).

Enoxacin hydrate is a miRNA processing activator and enhances siRNA-mediated mRNA degradation and promotes the biogenesis of endogenous miRNAs. Enoxacin hydrate has potent activities against gram-positive and -negative bacteria. Enoxacin hydrate is a cancer-specific growth inhibitor that acts by enhancing TAR RNA-binding protein 2 (TRBP)-mediated microRNA processing.

Specifications

Chemical Name or Material Enoxacin hydrate
CAS 84294-96-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C15H17FN4O3.3/2H2O
Quantity 500 mg
Synonym Enoxacin sesquihydrateAT-2266 hydrateCI-919 hydrate
Solubility Information 1M NaOH : 100 mg/mL (287.90 mM; ultrasonic and adjust pH to 11 with NaOH) ∣DMSO : 2.78 mg/mL (8.00 mM; Need ultrasonic) ∣H2O : 1 mg/mL (2.88 mM; ultrasonic and warming and heat to 80°C)
SMILES O=C(C1=CN(CC)C2=C(C=C(F)C(N3CCNCC3)=N2)C1=O)O.[1.5H2O]
Molecular Weight (g/mol) 347.34
Formula Weight 347.34
Percent Purity 98.15%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.