Learn More
Invitrogen™ ENOX1 Polyclonal Antibody
GREENER_CHOICE
Product Code. 16500542
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16500542 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16500542 Supplier Invitrogen™ Supplier No. PA597708
Please to purchase this item. Need a web account? Register with us today!
Rabbit Polyclonal Antibody
ENOX1 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Probably acts as a terminal oxidase of plasma electron transport from cytosolic NAD(P)H via hydroquinones to acceptors at the cell surface. Hydroquinone oxidase activity alternates with a protein disulfide-thiol interchange/oxidoreductase activity which may control physical membrane displacements associated with vesicle budding or cell enlargement. The activities oscillate with a period length of 24 minutes and play a role in control of the ultradian cellular biological clock.
TRUSTED_SUSTAINABILITY
Specifications
Antigen ENOX1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene ENOX1
Gene Accession No. Q8TC92
Gene Alias bA64J21.1; candidate growth-related and time keeping constitutive hydroquinone (NADH) oxidase; candidate growth-related and time keeping constitutive hydroquinone [NADH] oxidase; cCNOX; Cell proliferation-inducing gene 38 protein; CNOX; Constitutive Ecto-NOX; ecto-NOX disulfide-thiol exchanger 1; ENOX1; Hydroquinone [NADH] oxidase; PIG38; Protein disulfide-thiol oxidoreductase
Gene Symbols ENOX1
Host Species Rabbit
Immunogen Recombinant Human Ecto-NOX disulfide-thiol exchanger 1 protein (101-340AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55068
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG