missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eNOS, p Ser1177 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05432
This item is not returnable.
View return policy
Description
eNOS Polyclonal antibody specifically detects eNOS in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| eNOS | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
| Constitutive NOS, EC 1.14.13.39, ECNOS, endothelial, Endothelial NOS, eNOScNOS, nitric oxide synthase 3 (endothelial cell), NOS type III, NOSIII | |
| A synthetic phosphorylated peptide around S1177 of human eNOS (NP_000594.2). TQSFS | |
| 100 μg | |
| Cancer, HIF Target Genes, Hypoxia, Neuroscience, Neurotransmission, Phospho Specific | |
| 4846 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction