Learn More
ENMD-2076, MedChemExpress
Product Code. 16485246
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16485246 100 mg 100mg
16477047 5 mg 5mg
16460657 50 mg 50mg
16404338 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16485246 Supplier Medchem Express Supplier No. HY10987A
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress ENMD-2076 is a multi-targeted kinase inhibitor with IC50s of 1.86, 14, 58.2, 15.9, 92.7, 70.8, 56.4 nM for Aurora A, Flt3, KDR/VEGFR2, Flt4/VEGFR3, FGFR1, FGFR2, Src, PDGFRα, respectively.
ENMD-2076 is a multi-targeted kinase inhibitor with IC50s of 1.86, 14, 58.2, 15.9, 92.7, 70.8, 56.4 nM for Aurora A, Flt3, KDR/VEGFR2, Flt4/VEGFR3, FGFR1, FGFR2, Src, PDGFRα, respectively.
Specifications
Chemical Name or Material ENMD-2076
Physical Form Solid
CAS 934353-76-1
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C21H25N7
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : ≥ 31 mg/mL (82.56 mM)
SMILES CC1=CC(NC2=NC(/C=C/C3=CC=CC=C3)=NC(N4CCN(CC4)C)=C2)=NN1
Molecular Weight (g/mol) 375.47
Formula Weight 375.47
Percent Purity 98.17%
Grade Research