missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Enkurin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100999
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total C10orf63.
Enkurin interacts with transient receptor potential canonical.
Specifications
| Enkurin | |
| Polyclonal | |
| Unconjugated | |
| ENKUR | |
| 4933434I06Rik; AU016229; C10orf63; CFAP106; Enkur; Enkurin; enkurin, TRPC channel interacting protein; RGD1309839 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 219670, 71233 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6SP97, Q8TC29 | |
| ENKUR | |
| A synthesized peptide derived from human ENKUR(Accession Q8TC29), corresponding to amino acid residues N109-C159. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction