Learn More
Karl Hecht™ Digital Interval Timer
Product Code. 10668471
Click to view available options
Time Range:
99 min. 59 sec.
Unit Size:
Each
Product Code. 10668471
Brand: Karl Hecht™ 41977030
Only null left
Add to basket
Add to basket
Quartz-controlled, multi-function timer. Karl Hecht™ Digital Interval Timer is designed with a crisp LCD display panel.
  • Silver exterior color.
  • LCD display: 4 digits showing minutes and seconds.
  • Programmable up to 99 minutes, 59 seconds.
  • Counts down in seconds.
  • Alarm sounds for 30 seconds.
  • Memory recall function.
  • Individual settings for minutes and seconds.
  • Magnetic back.
  • Includes battery.
Specifications
Time Range 99 min. 59 sec.
Alarms For 30 sec.
Display LCD
Battery Type LR44
Includes Magnetic back