Learn More
Enecadin, MedChemExpress
Product Code. 16430509
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16430509 1 mg 1mg
16415527 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16430509 Supplier Medchem Express Supplier No. HY100119
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
MedChemExpress Enecadin is a neuroprotective agent extracted from patent US 8623823 B2.
Enecadin is a neuroprotective agent extracted from patent US 8623823 B2.
Specifications
Chemical Name or Material Enecadin
CAS 259525-01-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C21H28FN3O
Quantity 1 mg
Solubility Information DMSO : ≥ 50 mg/mL (139.88 mM)
SMILES CC1=NC(OCCCCCN2CCCCC2)=CC(C3=CC=C(F)C=C3)=N1
Molecular Weight (g/mol) 357.46
Formula Weight 357.46
Percent Purity 99.71%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation