missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Endoxifen Z-isomer hydrochloride, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
Description
Endoxifen Z-isomer hydrochloride is the most important Tamoxifen metabolite responsible for eliciting the anti-estrogenic effects of this drug in breast cancer cells expressing estrogen receptor-alpha (ERα).
Specifications
Specifications
| Chemical Name or Material | Endoxifen Z-isomer hydrochloride |
| CAS | 1032008-74-4 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C25H28ClNO2 |
| Quantity | 10 mg |
| Solubility Information | DMSO : 50 mg/mL (121.97 mM; Need ultrasonic) |
| SMILES | OC1=CC=C(/C(C2=CC=C(OCCNC)C=C2)=C(C3=CC=CC=C3)\CC)C=C1.[H]Cl.[Z] |
| Molecular Weight (g/mol) | 409.95 |
| Formula Weight | 409.95 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction