Learn More
MedChemExpress Endothelin 1 (swine, human)
Specifications
Specifications
| Purity | 95.44% |
| CAS | 117399-94-7 |
| Content And Storage | Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month |
| For Use With (Application) | COVID-19-immunoregulation |
| Molecular Weight (g/mol) | 2491.9 g/mol |
| Product Type | Reference compound |
| Quantity | 10 mg |
| Shipping Condition | Room temperature |
| Solubility | H2O : 1.82 mg/mL (0.73 mM∣ultrasonic and adjust pH to 3 with HCl) |