Learn More
Invitrogen™ Endothelin 1 Monoclonal Antibody (3G10)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA183134
Description
For IHC applications, frozen sections processed with dry acetone are recommended. Paraformaldehyde is not recommended as it destroys the epitope. Rat anti Human endothelin antibody, clone 3G10 reacts with the C terminal of endothelins 1, 2 and 3.
The protein encoded by this gene is proteolytically processed to release a secreted peptide termed endothelin 1. This peptide is a potent vasoconstrictor and is produced by vascular endothelial cells. Endothelin 1 also can affect the central nervous system. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
| Endothelin 1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| P05305 | |
| EDN1 | |
| Synthetic peptide corresponding to the c-terminal region of human endothelin (C,H,L,D,I,I,W), conjugated to BSA. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Frozen), Immunoradiometric Assay | |
| 3G10 | |
| Unconjugated | |
| EDN1 | |
| ARCND3; Big endothelin 1; Big endothelin-1; Big ET; Big ET 1; Big ET1; BigET; EDN1; endothelin 1; endothelin-1; Et; ET1; ET-1; HDLCQ7; PPET1; ppET-1; preproendothelin; preproendothelin-1; preproET; QME | |
| Rat | |
| Protein G | |
| RUO | |
| 1906 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.