Learn More
Endo Lurasidone Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | Endo Lurasidone Hydrochloride |
| CAS | 1318074-25-7 |
| Molecular Formula | C28H36N4O2S . HCl |
| Quantity | 50 mg |
| Synonym | (3aR,4R,7S,7aS)-2-(((1R,2R)-2-((4-(Benzo[d]isothiazol-3-yl)piperazin-1-yl)methyl)cyclohexyl)methyl)hexahydro-1H-4,7-methanoisoindole-1,3(2H)-dione Hydrochloride |
| InChI Formula | InChI=1S/C28H36N4O2S.ClH/c33-27-24-18-9-10-19(15-18)25(24)28(34)32(27)17-21-6-2-1-5-20(21)16-30-11-13-31(14-12-30)26-22-7-3-4-8-23(22)35-29-26;/h3-4,7-8,18-21,24-25H,1-2,5-6,9-17H2;1H/t18-,19+,20-,21-,24-,25+;/m0./s1 |
| SMILES | Cl.O=C1[C@@H]2[C@@H]3CC[C@@H](C3)[C@@H]2C(=O)N1C[C@@H]4CCCC[C@H]4CN5CCN(CC5)c6nsc7ccccc67 |
| Molecular Weight (g/mol) | 529.1 |