Learn More
EMILIN3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA563792
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: SPHGRKGPGL FGERLERLEG DVQRLAQTYG TLSGLVASHE DPNRMTGGPR APAVPVGFGV IPEGLVGPGD RARGPLTPPL DEILSKVTEV SNTLQT Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 77%.
EMILIN3 Polyclonal antibody specifically detects EMILIN3 in Human samples. It is validated for Immunohistochemistry.Specifications
EMILIN3 | |
Polyclonal | |
Unconjugated | |
EMILIN3 | |
C20orf130; dJ620E11.4; Elastin microfibril interface-located protein 3; elastin microfibril interface-located protein 5; Elastin microfibril interfacer 3; elastin microfibril interfacer 5; EMILIN3; EMILIN-3; EMILIN5; EMILIN-5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
90187 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NT22 | |
EMILIN3 | |
Recombinant protein corresponding to Human EMILIN3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only