Specifications
| For Use With (Equipment) | 12 x 75 mm Tubes |
| For Use With (Application) | Radionucleotide counting by transfer of individual filters to tubes |
| Includes | Multiple punch, 1 carrier rack for 12 x 75mm tubes |
| Quantity | 1/Pk. |