Learn More
Merck MultiScreen™ Column Loaders
Product Code. 10760113 Shop All Merck Products
Click to view available options
Volume (Metric) Well:
25 μL
45 μL
100 μL
Unit Size:
Each
Product Code. 10760113
Brand: Merck MACL09625
Please to purchase this item. Need a web account? Register with us today!
No need to pipet slurries, gels, or resins; load all 96 wells simultaneously and uniformly
  • Constructed of aluminum with an abrasion-resistant, chemically inert, nonstick coating
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Array 8 x 12
Color Black
Volume (Metric) Well 25 μL
No. of Wells 96
Sterility Non-sterile
For Use With (Application) Load 25μL resin volumes simultaneously into 96 wells of MultiScreen plate for resin or bead based assays, chromatography
Quantity 1/Pk.