Learn More
Merck Millipore M Air T™ Agar Cassettes
Ready-to-use cassettes, prefilled with gamma-sterilized medium
Description
- Fast and reliable results with maximum recovery
- Large agar amount and unique micro-perforated sieve allow air sampling of up to 1 cubic meter
- Unit-to-unit consistency of agar surface shape
- D
- Reproducible results
Compliance
USP