missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EMC7 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15998214
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15998214 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15998214 Supplier Invitrogen™ Supplier No. PA565474

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: VVPGVKPQDWI SAARVLVDGE EHVGFLKTDG SFVVHDIPSG SYVVEVVSPA YRFDPVRVDI TSKGKMRARY VNYIKTSEVV RLPYPLQMKS SGPPSYFIKR ESWGW Highest antigen sequence identity to the following orthologs - mouse 98%, rat 98%.

EMC7 (ER Membrane Protein Complex Subunit 7) is a Protein Coding gene. Diseases associated with EMC7 include Cone-Rod Dystrophy, X-Linked, 1. Gene Ontology (GO) annotations related to this gene include carbohydrate binding. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen EMC7
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EMC7
Gene Accession No. Q9NPA0
Gene Alias 2900064A13Rik; AI451465; c11orf3; C15orf24; chromosome 15 hypothetical ATG/GTP binding protein; Emc7; ER membrane protein complex subunit 7; HT022; ORF1-FL1; Orf3; UNQ905/PRO1926; UPF0480 protein C15orf24; UPF0480 protein C15orf24 homolog
Gene Symbols EMC7
Host Species Rabbit
Immunogen Recombinant Human EMC7. Recombinant protein control fragment (Product #RP-106150).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56851
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.