missing translation for 'onlineSavingsMsg'
Learn More

Embelin, MedChemExpress

Product Code. 16423387
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16423387

Brand: Medchem Express HY17473

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Embelin (Embelic acid), a potent, nonpeptidic XIAP inhibitor (IC50=4.1 μM), inhibits cell growth, induces apoptosis, and activates caspase-9 in prostate cancer cells with high levels of XIAP. Embelin blocks NF-kappaB signaling pathway leading to suppression of NF-kappaB-regulated antiapoptotic and metastatic gene products. Embelin also induces autophagic and apoptotic cell death in human oral squamous cell carcinoma cells.

Embelin (Embelic acid), a potent, nonpeptidic XIAP inhibitor (IC50=4.1 μM), inhibits cell growth, induces apoptosis, and activates caspase-9 in prostate cancer cells with high levels of XIAP. Embelin blocks NF-kappaB signaling pathway leading to suppression of NF-kappaB-regulated antiapoptotic and metastatic gene products. Embelin also induces autophagic and apoptotic cell death in human oral squamous cell carcinoma cells.

Specifications

Chemical Name or Material Embelin
Physical Form Solid
CAS 550-24-3
Color Orange
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C17H26O4
For Use With (Application) Cancer-programmed cell death
Synonym Embelic acid Emberine NSC 91874
Solubility Information DMSO : ≥ 50 mg/mL (169.84 mM)
SMILES O=C1C(O)=C(CCCCCCCCCCC)C(C(O)=C1)=O
Molecular Weight (g/mol) 294.39
Formula Weight 294.39
Percent Purity 98.19%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.