missing translation for 'onlineSavingsMsg'
Learn More

EMAP-II/AIMP1 Antibody (959516) [FITC], Novus Biologicals™

Product Code. 30671404 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30671404 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30671404 Supplier Novus Biologicals Supplier No. FAB1910F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

EMAP-II/AIMP1 Monoclonal antibody specifically detects EMAP-II/AIMP1 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen EMAP-II/AIMP1
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 959516
Conjugate FITC
Formulation PBS
Gene Alias aminoacyl tRNA synthetase complex-interacting multifunctional protein 1, EMAP-2, EMAPIImember 1 (endothelialmonocyte-activating), endothelial-monocyte activating polypeptide II, Multisynthase complex auxiliary component p43, multisynthetase complex auxiliary component p43
Host Species Mouse
Immunogen E.coli-derived recombinant human EMAP-II, Ser147-Lys312, Accession # Q12904
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 9255
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.