Learn More
Tocris Bioscience™ EM12-SO2F
Description
EM12-SO2F is a potent covalent Cereblon (CRBN) inhibitor (IC50 = 0.88 nM). Binds to a histidine residue (His353) in the IMiD binding pocket of CRBN and blocks the recruitment of its substrates. EM12-SO2F prevents Lenalidomide (Cat. No. 6305)-induced degradation of the zinc finger transcription factor IKZF1 in MOLT4 cells. It complements CRBN based PROTAC and genetic methods to validate CRBN-mediated degradation.
Specifications
Specifications
| Chemical Name or Material | 2-(2,6-Dioxo-3-piperidinyl)-2,3-dihydro-3-oxo-1H-isoindole-5-sulfonyl fluoride |
| CAS | 2803819-61-4 |
| Quantity | 10 mg |
| Target | CRBN |
| Molecular Formula | C13H11FN2O5S |
| Purity | 95% |