Learn More
ELP2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA531859
Additional Details : Weight : 0.01000kg
Description
PA5-31859 targets ELP2 in IF, IHC (P), and WB applications and shows reactivity with Human samples. The PA5-31859 immunogen is recombit fragment corresponding to a region within amino acids 318 and 620 of Human ELP2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
Regulates the ligand-dependent activation of STAT3. Acts as subunit of the RNA polymerase II elongator complex, which is a histone acetyltransferase component of the RNA polymerase II (Pol II) holoenzyme and is involved in transcriptional elongation. Elongator may play a role in chromatin remodeling and is involved in acetylation of histones H3 and probably H4.Specifications
ELP2 | |
Polyclonal | |
Unconjugated | |
Q6IA86 | |
ELP2 | |
Recombit fragment corresponding to a region within amino acids 318 and 620 of Human ELP2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1 mg/mL | |
ELP2 | |
SHINC-2, STATIP1, StIP, STAT3-interacting protein 1, elongator complex protein 2, elongator protein 2, signal transducer and activator of transcription 3 interacting protein 1, signal transducer and activator of transcription interacting protein 1, stIP1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55250 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |