missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ELOVL2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 13260289
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13260289 400 μL 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13260289 Supplier Invitrogen™ Supplier No. PA526199

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody is predicted to react with mouse based on sequence homology.

ELOVL2 could be implicated in tissue-specific synthesis of very long chain fatty acids and sphingolipids. This protein may catalyze one or both of the reduction reaction in fatty acid elongation, i. e., conversion of beta-ketoacyl CoA to beta-hydroxyacyl CoA or reduction of trans-2-enoyl CoA to the saturated acyl CoA derivative.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ELOVL2
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ELOVL2
Gene Accession No. Q9NXB9
Gene Alias 3-keto acyl-CoA synthase ELOVL2; AI317360; ELG3; elongation of very long chain fatty acids (FEN1/Elo2, SUR4/Elo3, yeast)-like 2; elongation of very long chain fatty acids protein 2; elongation of very long chain fatty acids-like 2; ELOVL FA elongase 2; ELOVL family member 2; ELOVL fatty acid elongase 2; Elovl2; sequence similarity to Cig30 2; SSC2; Very long chain 3-ketoacyl-CoA synthase 2; very long chain 3-oxoacyl-CoA synthase 2; very-long-chain 3-oxoacyl-CoA synthase 2
Gene Symbols ELOVL2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 1-27 amino acids from the N-terminal region of human ELOVL2.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54898
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.