Learn More
ELL2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA564949
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: HLTNRVPPTLN GHLNPTSEKS AAGLPLPPAA AAIPTPPPLP STYLPISHPP QIVNSNSNSP STPEGRGTQD LPVDSFSQND SIYEDQQDKY TSRTSLETLP PGSVLLKCPK PMEENHS Highest antigen sequence identity to the following orthologs - mouse 84%, rat 83%.
ELL2 is the elongation factor that can increase the catalytic rate of RNA polymerase II transcription by suppressing transient pausing by the polymerase at multiple sites along the DNA.Specifications
ELL2 | |
Polyclonal | |
Unconjugated | |
ELL2 | |
RNA polymerase II elongation factor ELL2; ELL-related RNA polymerase II, elongation factor; elongation factor RNA for polymerase II 2; elongation factor, RNA polymerase II, 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22936 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O00472 | |
ELL2 | |
Recombit Human ELL2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only