Learn More
Invitrogen™ ELL2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100683
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ELL2.
SHP-1 (PTPN6) is a non-receptor protein tyrosine phosphatase that is expressed primarily in hematopoietic cells. The enzyme is composed of two SH2 domains, a tyrosine phosphatase catalytic domain, and a carboxy-terminal regulatory domain. SHP-1 removes phosphates from target proteins to downregulate several tyrosine kinase-regulated pathways. In hematopoietic cells, the amino-terminal SH2 domain of SHP-1 binds to tyrosine phosphorylated erythropoietin receptors (EpoR) to negatively regulate hematopoietic growth. Overexpression of SHP-1 in epithelial cells results in dephosphorylation of the Ros receptor tyrosine kinase and subsequent downregulation of Ros-dependent cell proliferation and transformation. Following ligand binding in myeloid cells, SHP-1 associates with the IL-3R beta chain and downregulates IL-3-induced tyrosine phosphorylation and cell proliferation.
Specifications
| ELL2 | |
| Polyclonal | |
| Unconjugated | |
| ELL2 | |
| cb244; eleven-nineteen lysine-rich leukemia gene 2; ELL2; ELL-related RNA polymerase II, elongation factor; elongation factor for RNA polymerase II 2; elongation factor RNA for polymerase II 2; elongation factor RNA polymerase II 2; elongation factor, RNA polymerase II 2; elongation factor, RNA polymerase II, 2; fb78f03; im:3814960; RNA polymerase II elongation factor ELL2; RNA polymerase II elongation factor ELL2-like protein; sb:cb244; wu:fb78f03; wu:fb78f03 protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 192657, 22936 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O00472, Q3UKU1 | |
| ELL2 | |
| A synthesized peptide derived from human ELL2(Accession O00472), corresponding to amino acid residues G161-I211. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |