missing translation for 'onlineSavingsMsg'
Learn More

Elinogrel, MedChemExpress

Product Code. p-200100553
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
This item is not returnable. View return policy

Product Code. 16491139

Brand: Medchem Express HY11021

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Elinogrel (PRT060128) is a potent, direct acting, competitive, and reversible platelet P2Y12 antagonist (IC50=20 nM). It is orally and intravenously available and has potent antiplatelet effects.

Elinogrel (PRT060128) is a potent, direct acting, competitive, and reversible platelet P2Y12 antagonist (IC50=20 nM). It is orally and intravenously available and has potent antiplatelet effects.

Specifications

Chemical Name or Material Elinogrel
CAS 936500-94-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C20H15ClFN5O5S2
Quantity 1 mg
Synonym PRT060128
Solubility Information DMSO : 100 mg/mL (190.86 mM; Need ultrasonic)
SMILES O=S(C1=CC=C(Cl)S1)(NC(NC2=CC=C(N3C(NC4=C(C=C(F)C(NC)=C4)C3=O)=O)C=C2)=O)=O
Molecular Weight (g/mol) 523.95
Formula Weight 523.95
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.