Learn More
Elinogrel, MedChemExpress
Product Code. 16491139
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Product Code. 16491139
Brand: Medchem Express HY11021
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Elinogrel (PRT060128) is a potent, direct acting, competitive, and reversible platelet P2Y12 antagonist (IC50=20 nM). It is orally and intravenously available and has potent antiplatelet effects.
Elinogrel (PRT060128) is a potent, direct acting, competitive, and reversible platelet P2Y12 antagonist (IC50=20 nM). It is orally and intravenously available and has potent antiplatelet effects.
Specifications
Chemical Name or Material Elinogrel
CAS 936500-94-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C20H15ClFN5O5S2
Quantity 1 mg
Synonym PRT060128
Solubility Information DMSO : 100 mg/mL (190.86 mM; Need ultrasonic)
SMILES O=S(C1=CC=C(Cl)S1)(NC(NC2=CC=C(N3C(NC4=C(C=C(F)C(NC)=C4)C3=O)=O)C=C2)=O)=O
Molecular Weight (g/mol) 523.95
Formula Weight 523.95
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation