Learn More
Karl Hecht™ Electronic Maximum-minimum Thermometer
Click to view available options
Display:
High/Low, LED
Unit Size:
Each
Description
- Highest/lowest temperature indicator displays.
- Switchable °C/°F .
- Hanging or standing options.
- Extremely thin cable facilitates threading through window seal, etc.
- Waterproof exterior sensor.
Specifications
Specifications
| For Use With (Application) | For measuring indoor and outdoor temperatures |
| Length (Metric) Cable | 1.2 m |
| Display | High/Low, LED |