missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELAC2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA541987
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: ITCNPEEFIV EALQLPNFQQ SVQEYRRSAQ DGPAPAEKRS QYPEIIFLGT Sequence homology: Dog: 100%; Human: 100%; Mouse: 83%; Rat: 83%
ELAC2 Polyclonal antibody specifically detects ELAC2 in Human samples. It is validated for Western Blot.Specifications
ELAC2 | |
Polyclonal | |
Unconjugated | |
Elac2 | |
1110017O07Rik; COXPD17; D11Wsu80e; elaC homolog 2; elaC homolog 2 (E. coli); elaC homolog protein 2; elaC ribonuclease Z 2; ELAC2; elaC-like protein 2; ELC2; heredity prostate cancer protein 2; HPC2; putative prostate cancer susceptibility protein HPC2/ELAC2; ribonuclease Z 2; RNase Z 2; tRNA 3 endonuclease 2; tRNase Z (long form); tRNase Z 2; tRNase Z(L); zinc phosphodiesterase ELAC protein 2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
60528 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9BQ52 | |
Elac2 | |
Synthetic peptide directed towards the C-terminal of human ELAC2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |