missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Triglyceride (TG) Colorimetric Assay Kit

Product Code. 17972071
Change view
Click to view available options
Quantity:
96 tests
Unit Size:
Pack of 96
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17972071 96 tests Pack of 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17972071 Supplier Invitrogen™ Supplier No. EEA028

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

These complete, ready-to-use kits include clear 96-well plate(s), other assay specific reagents, buffers, and a lot-specific technical data sheet.

The Triglyceride (TG) Colormetric Assay Kit is a highly reliable and sensitive assay designed for the quantitative measurement of triglycerides in serum, plasma, and tissue samples. Triglycerides are a type of lipid molecule that play a crucial role in energy storage and metabolism. They are the main component of vegetable oil, animal fat, low-density lipoprotein (LDL), and very low-density lipoprotein (VLDL). Elevated levels of triglycerides in the blood have been associated with an increased risk of cardiovascular disease, glucose metabolism, insulin resistance, and Type 2 diabetes.

The assay utilizes the GPO-PAP method, which involves the hydrolysis of triglycerides to glycerol and fatty acids. Glycerol is then oxidized by glycerol kinase to glycerol-3-phosphate, which is oxidized by GPO and produces hydrogen peroxide. In the presence of 4-aminoantipyrine and phenol, hydrogen peroxide is catalyzed by peroxidase (POD) to produce colored quinones which can be measured at 510 nm to calculate the amount of TG in the sample.

Key features

  • Simple and fast protocol—user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
  • Broad range of kits—across key analytes and targets within the most important metabolic pathways
  • Consistent—strict quality control criteria to ensure reproducible and reliable results
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.14–10 mmol/L
Assay Sensitivity 0.14 mmol/L
Content And Storage Store at 2°C to 8°C.
Detection Method Colorimetric
Sample Type Serum, Plasma, Tissue
For Use With (Equipment) Microplate Reader
Incubation Time 10 min.
Interassay CV 9.2
Intraassay CV 4.1
Quantity 96 tests
Sample Volume 2.5 μL
Shipping Condition Dry Ice, 2–8°C
Target Kit Named Triglyceride (TG)
Total Assay Time 30 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.