Learn More
Tocris Bioscience™ eIF4A i28
Description
eIF4A i28 is a eukaryotic initiation factor-4A (eIF4A) inhibitor (IC50 = 8.60 μM in an eIF4A ATPase assay). It decreases BJAB Burkitt lymphoma cell viability (EC50 = 0.46 μM in a CellTiter-Glo (CTG) luminescence cell viability assay. It acts as an RNA-competitive, ATP-uncompetitive eIF4A inhibitor.
Specifications
Specifications
| Chemical Name or Material | 2-(5-(4-Butylphenyl)furan-2-yl)-6-nitroquinoline-4-carboxylic acid |
| CAS | 2861994-92-3 |
| Quantity | 10 mg |
| Target | DNA, RNA and Protein Synthesis Inhibitors |
| Molecular Formula | C24H20N2O5 |
| Purity | 0.98 |