Learn More
Tocris Bioscience™ EIDD 1931
Description
EIDD 1931 is a viral RNA-dependent RNA polymerase (RdRP) inhibitor and broad spectrum antiviral nucleotide. Inhibits replication of a range of RNA viruses including influenza, Ebola, HCV, and human and zoonotic coronaviruses (IC50 values are 0.15 and 0.3 μM for inhibition of MERS-CoV and SARS-CoV-2 replication in vitro). Increases mutation rates and induces lethal mutagenesis in viral RNA. Also inhibits replication of mouse hepatitis virus with mutations conferring resistance to remdesivir (Cat. No. 7226).
Specifications
Specifications
| Chemical Name or Material | N-Hydroxycytidine |
| CAS | 3258-02-4. |
| Quantity | 25 mg |
| Target | RNA/DNA Polymerase Inhibitors |
| Molecular Formula | C9H13N3O6 |
| Purity | 0.98 |