Learn More
Eicosapentaenoic Acid, MedChemExpress
Product Code. p-200096362
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
Product Code. 16494437
Brand: Medchem Express HYB0660
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Eicosapentaenoic Acid (EPA) is an orally active Omega-3 long-chain polyunsaturated fatty acid (ω-3 LC-PUFA).
Eicosapentaenoic Acid exhibits a DNA demethylating action that promotes the re-expression of the tumor suppressor gene CCAAT/enhancer-binding protein δ (C/EBPδ). Eicosapentaenoic Acid activates RAS/ERK/C/EBPβ pathway through H-Ras intron 1 CpG island demethylation in U937 leukemia cells. Eicosapentaenoic Acid can promote relaxation of vascular smooth muscle cells and vasodilation.
Specifications
Chemical Name or Material Eicosapentaenoic Acid
CAS 10417-94-4
Color Colorless
Purity Grade Notes Research
Molecular Formula C20H30O2
Quantity 50 mg
Synonym EPA Timnodonic acid
Solubility Information Ethanol : ≥ 100 mg/mL (330.63 mM) ∣DMSO : ≥ 53 mg/mL (175.24 mM)
SMILES CC/C=C\C/C=C\C/C=C\C/C=C\C/C=C\CCCC(O)=O
Molecular Weight (g/mol) 302.45
Formula Weight 302.45
Percent Purity 98.0%
Grade Research
Physical Form Liquid
For Use With (Application) Cancer-programmed cell death