missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EGR1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599273
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total EGR1.
The protein encoded by this gene belongs to the EGR family of C2H2-type zinc-finger proteins. It is a nuclear protein and functions as a transcriptional regulator. The products of target genes it activates are required for differentitation and mitogenesis. Studies suggest this is a cancer suppresor gene.
Specifications
| EGR1 | |
| Polyclonal | |
| Unconjugated | |
| EGR1 | |
| A530045N19Rik; AT225; early growth response 1; early growth response protein 1; egr; EGR1; EGR-1; ETR103; G0S30; Krox-1; Krox24; Krox-24; Krox-24 nuclear protein; Nerve growth factor-induced protein A; Ngf1; NGF1-A; Ngfi; NGFIA; NGFI-A; TIS8; transcription factor ETR103; Transcription factor Zif268; Zenk; Zfp-6; Zif268; zif-268; Zinc finger protein 225; zinc finger protein Krox-24; ZNF225 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 13653, 1958 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P08046, P18146 | |
| EGR1 | |
| A synthesized peptide derived from human EGR1(Accession P18146), corresponding to amino acid residues S441-P461. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction