missing translation for 'onlineSavingsMsg'
Learn More

eGFP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

265.00 € - 913.00 €

Specifications

Antigen eGFP
Dilution ELISA 1:5000-1:10000
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18698956
View Documents
Novus Biologicals
NBP3-06640-100ul
100 μg
419.00 €
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18612918
View Documents
Novus Biologicals
NBP3-06640-500ul
500 μg
913.00 €
500µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18613288
View Documents
Novus Biologicals
NBP3-06640-50ul
50 μg
265.00 €
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

eGFP Polyclonal antibody specifically detects eGFP in Nonspecies specific samples. It is validated for ELISA
Specifications

Specifications

eGFP
ELISA
Unconjugated
Rabbit
Non-species specific
Enhanced Green Fluorescent Protein, GFP, Green Fluorescent Protein
IgG
Antigen and protein A Affinity-purified
ELISA 1:5000-1:10000
Polyclonal
Purified
RUO
0.2 um filtered solution in PBS
Produced in rabbits immunized with purified, recombinant eGFP (AFI57842.2; Met1-Lys239).
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.