Learn More
Eganelisib, MedChemExpress
Product Code. 16483128
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16483128
Brand: Medchem Express HY10071610mg
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Eganelisib (IPI549) is a potent and selective PI3Kγ inhibitor with an IC50 of 16 nM. Eganelisib shows >100-fold selectivity over other lipid and protein kinases.
Eganelisib (IPI549) is a potent and selective PI3Kγ inhibitor with an IC50 of 16 nM. Eganelisib shows >100-fold selectivity over other lipid and protein kinases.
Specifications
Chemical Name or Material Eganelisib
Physical Form Powder
CAS 1693758-51-8
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C30H24N8O2
For Use With (Application) Cancer-Kinase/protease
Synonym IPI-549
Solubility Information DMSO : 15 mg/mL (28.38 mM; Need ultrasonic and warming)
SMILES O=C(C1=C2N=CC=CN2N=C1N)N[C@H](C3=CC4=C(C(N3C5=CC=CC=C5)=O)C(C#CC6=CN(C)N=C6)=CC=C4)C
Molecular Weight (g/mol) 528.56
Formula Weight 528.56
Percent Purity 95.0%
Grade Research